Skip to Content
Merck

76172BC

MVP ICON® Buffer Solution, pH 4.0 Red

for use with MVP ICON® pH Probe (78088BC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122409
NACRES:
NA.25
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

pH calibration

Quality Segment

product line

MVP ICON®

form

liquid

manufacturer/tradename

MVP ICON®

application(s)

food and beverages

compatibility

MVP ICON®, for use with MVP ICON® pH Probe (78088BC)

General description

pH 4.0 Red Buffer Solution (500 mL) for the MVP ICON® pH Probe

Application

For use in performing 3-point calibration of the MVP ICON® pH/Temperature Probe (78088BC)

Legal Information

MVP ICON is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library