Skip to Content
Merck
All Photos(1)

Key Documents

69967-M

Sigma-Aldrich

pT7Blue DNA (uncut) - Novagen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106610
NACRES:
NA.51

Pricing and availability is not currently available.

form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−20°C

General description

pT7Blue contains the pUC19 backbone (including high-copy number origin of replication and lac sequences), a T7 promoter, f1 origin of replication, and modified multiple cloning region. The multiple cloning region contains an EcoR V site used for T-cloning flanked by an Nde I site, which allows inserts to be conveniently subcloned into the Nde I site of many pET vectors.

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PT7BLUE is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service