Skip to Content
Merck
All Photos(1)

Documents

402088

Sigma-Aldrich

Indirubin-3′-monoxime-5-sulphonic Acid

A potent, reversible, and selective inhibitor of Cdk1 (IC₅₀ = 5 nM) and Cdk5 (IC₅₀ = 7 nM) and glycogen synthase kinase-3β (GSK-3β; IC₅₀ = 80 nM).

Synonym(s):

Indirubin-3′-monoxime-5-sulphonic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11N3O5S
CAS Number:
Molecular Weight:
357.34
UNSPSC Code:
12352106
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark red

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C16H11N3O5S/c20-16-13(10-7-8(25(22,23)24)5-6-12(10)18-16)15-14(19-21)9-3-1-2-4-11(9)17-15/h1-7,17,19,21H,(H,22,23,24)

InChI key

IZDNACYDKBNHHC-UHFFFAOYSA-N

General description

A highly potent, reversible, and selective inhibitor of cyclin-dependent kinases 1 and 5 (IC50 = 5 nM for CDK1; IC50 = 7 nM for CDK5). Reported to act by competing with ATP for binding to the catalytic site of the kinase. Also acts as a potent inhibitor of glycogen synthase kinase-3β (GSK-3β; IC50 = 80 nM).
A potent, reversible, and selective inhibitor of Cdk1 (IC50 = 5 nM) and Cdk5 (IC50 = 7 nM) and glycogen synthase kinase-3β (GSK-3β; IC50 = 80 nM). The inhibition is competitive with respect to ATP.

Biochem/physiol Actions

Cell permeable: no
Primary Target
CDK1
Product competes with ATP.
Reversible: yes
Target IC50: 5 nM, 7 nM, 80 nM, against Cdk1, Cdk5, and glycogen synthase kinase-3β (GSK-3β), respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze at -20°C. Stock solutions are stable for up to 4 months at -20°C.

Other Notes

Leclerc, S., et al. 2001. J. Biol. Chem.276, 251.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service