Skip to Content
Merck
All Photos(1)

Documents

1.46413

Millipore

Malachite Green Broth

Application: Selective enrichment of Pseudomonas aeruginosa
Packaging: 50 ml in 275 ml wide mouth bottle (6 bottles per box)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106226
NACRES:
NA.74

Quality Level

form

liquid

feature

ready-to-use

pH

7.3 ( in H2O)

application(s)

food and beverages
industrial qc
microbiology

storage temp.

2-25°C

suitability

Pseudomonas spp.

General description

Application: Selective enrichment of Pseudomonas aeruginosa

Packaging: 50 ml in 275 ml wide mouth bottle (6 bottles per box)

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service