Skip to Content
Merck
All Photos(1)

Key Documents

1.16819

Millipore

Anaerocult IS

suitable for microbiology, Gas generator system for the anaerobic incubation of identification systems and sensitivity tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104308

form

dry powder (dehydrated (DCM))

manufacturer/tradename

Anaerocult

bulk density

550 kg/m3

suitability

suitable for microbiology
anaerobic bacteria

application(s)

food and beverages
microbiology

storage temp.

−20°C

General description

ANAEROCULT IS contains components that chemically bind any oxygen present within a short space of time and release carbon dioxide thus creating an anaerobic atmosphere. The special incubation bag is designed to accommodate all identification galleries currently on the market.
for microbiology Gas generator system for the anaerobic incubation of identification systems and sensitivity tests

Application

ANAEROCULT IS gas generating system is used for the anaerobic incubation of identification systems and susceptibility test. Each item for incubation can be evaluated without disrupting the anaerobic conditions. Results that are initially unclear can be checked repeatedly without having to open the incubation bag.

Analysis Note

Growth
- Bacteroides fragilis ATCC 25285
passes test - Clostridium beijerinckii ATCC 17795passes test

Legal Information

ANAEROCULT is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C M Braun et al.
Biochemical pharmacology, 56(7), 871-879 (1998-10-17)
While a differential sensitivity to cyclic AMP (cAMP)-mediated signaling between Th1 and Th2 cells has been hypothesized, differential activity of downstream signaling through cAMP-dependent protein kinase (cAK) isoforms remains unexplored. We herein report the effects of type 1- and type
H Yokozaki et al.
Cancer research, 52(9), 2504-2508 (1992-05-01)
A set of adenosine 3':5'-monophosphate (cAMP) analogues that combine exocyclic sulfur substitutions in the equatorial (Rp) or the axial (Sp) position of the cyclophosphate ring with modifications in the adenine base of cAMP were tested for their effect on the
B T Gjertsen et al.
The Journal of biological chemistry, 270(35), 20599-20607 (1995-09-01)
Novel (Rp)-cAMPS analogs differed widely in ability to antagonize cAMP activation of pure cAMP-dependent protein kinase I and II and to antagonize actions of cAMP on gene expression, shape change, apoptosis, DNA replication, and protein phosphorylation in intact cells. These

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service