Skip to Content
Merck
All Photos(1)

Key Documents

1.02427

Supelco

1-Chloro-2,4-dinitrobenzene

GR for analysis

Synonym(s):

1-Chloro-2,4-dinitrobenzene, 4-Chloro-1,3-dinitrobenzene, 1,3-Dinitro-4-chlorobenzene, 2,4-Dinitro-1-chlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3ClN2O4
CAS Number:
Molecular Weight:
202.55
MDL number:
UNSPSC Code:
12352101
EC Index Number:
202-551-4

vapor pressure

1 hPa ( 106 °C)

Quality Level

Assay

≥99% (HPLC)

form

solid

autoignition temp.

464 °C

potency

640 mg/kg LD50, oral (Rat)
130 mg/kg LD50, skin (Rabbit)

expl. lim.

1.9-22 % (v/v)

bp

315 °C/1013 hPa

mp

48-52 °C

transition temp

flash point 194 °C

solubility

0.36 g/L

density

1.7 g/cm3 at 20 °C

bulk density

760 kg/m3

storage temp.

15-25°C

InChI

1S/C6H3ClN2O4/c7-5-2-1-4(8(10)11)3-6(5)9(12)13/h1-3H

InChI key

VYZAHLCBVHPDDF-UHFFFAOYSA-N

Analysis Note

Purity (HPLC): ≥ 99
Identity (IR-spectrum): passes test
Melting point: 49 - 52
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
Sulfated ash: ≤ 0.1 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

381.2 °F - closed cup

Flash Point(C)

194 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service