Skip to Content
Merck
All Photos(2)

Key Documents

07-593

Sigma-Aldrich

Anti-acetyl-Histone H3 (Lys9/18) Antibody

serum, Upstate®

Synonym(s):

H3K9/18Ac, Histone H3 (acetyl K9/18)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, Saccharomyces cerevisiae, bovine, human

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable
multiplexing: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys9/Lys18)

Gene Information

human ... H3F3B(3021)

General description

Histone H3 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure.

Acetylation of histone H3 occurs at several different lysine positions in the histone tail and is performed by a family of enzymes known as Histone Acetyl Transferases (HATs).

Specificity

Broad species cross-reactivity expected
Recognizes Histone H3 acetylated on lysines 9 and 18.

Immunogen

peptide (ARAcKSTGGKAPRAcKQL) which corresponds to acetyl-lysine at residue 9 and 18 of human Histone H3

Application

Anti-acetyl-Histone H3 (Lys9/18) Antibody is a rabbit polyclonal antibody for detection of acetyl-Histone H3 (Lys9/18) also known as H3K9/18Ac, Histone H3 (acetyl K9/18) & has been validated in ChIP, WB, Mplex.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

routinely evaluated by immunoblot on acid extracts from sodium butyrate treated Hela cells (Catalog #17-305)

Target description

17 kDa

Physical form

Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide as a preservative.
Unpurified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Acid extracts from sodium butyrate treated HeLa cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Site-specific loss of acetylation upon phosphorylation of histone H3
Edmondson, Diane G, et al
The Journal of Biological Chemistry, 277, 29496-29502 (2002)
Preliminary study of sperm chromatin characteristics of the brachyuran crab Maja brachydactyla. Histones and nucleosome-like structures in decapod crustacean sperm nuclei previously described without SNBPs.
K Kurtz, J Ausio, M Chiva, K Kurtz, J Ausio, M Chiva
Tissue & cell null
Michal Mikula et al.
International journal of molecular medicine, 34(6), 1647-1654 (2014-10-17)
Obesity contributes to the development of non-alcoholic fatty liver disease (NAFLD), which is characterized by the upregulated expression of two key inflammatory mediators: tumor necrosis factor (Tnfa) and monocyte chemotactic protein 1 (Mcp1; also known as Ccl2). However, the chromatin
Melatonin induces histone hyperacetylation in the rat brain.
Niles, Lennard P, et al.
Neuroscience Letters (2013)
Vijay S Thakur et al.
Carcinogenesis, 33(2), 377-384 (2011-11-25)
Green tea polyphenols (GTPs) reactivate epigenetically silenced genes in cancer cells and trigger cell cycle arrest and apoptosis; however, the mechanisms whereby these effects occur are not well understood. We investigated the molecular mechanisms underlying the antiproliferative effects of GTP

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service