Skip to Content
Merck
All Photos(4)

Key Documents

07-537

Sigma-Aldrich

Anti-Prox1 Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Prox1(19130)

Specificity

Prox1

Immunogen

KLH-conjugated, synthetic peptide (C-QSVSPRESYRENKRKQKLPQ) corresponding to amino acids 196-215 of human prospero-related homeobox 1 (Prox1) with an N-terminal cysteine added for conjugation purposes.

Application

Detect Prox1 using this Anti-Prox1 Antibody validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Sensory & PNS

Developmental Neuroscience

Quality

routinely evaluated by immunoblot in RIPA lysates from mouse liver cells

Target description

95kDa

Linkage

Replaces: MABE488

Physical form

0.05% sodium azide and 30% glycerol
ImmunoAffinity Purified

Storage and Stability

1 year at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mei-Ren Pan et al.
Journal of cell science, 122(Pt 18), 3358-3364 (2009-08-27)
Prox1 is a master regulator for the development of lymphatic vasculature and the induction of lymphangiogenesis. In this study, we identified Prox1 as a new target for small ubiquitin-like modifier 1 (SUMO-1). Lysine 556 (K556) was found to be the
Estrogen receptor ? induces antiinflammatory and antitumorigenic networks in colon cancer cells.
Edvardsson, K; Strom, A; Jonsson, P; Gustafsson, J?; Williams, C
Molecular Endocrinology null
Janina Becker et al.
Nature structural & molecular biology, 20(4), 525-531 (2013-03-19)
SUMOylation is an essential modification that regulates hundreds of proteins in eukaryotic cells. Owing to its dynamic nature and low steady-state levels, endogenous SUMOylation is challenging to detect. Here, we present a method that allows efficient enrichment and identification of
Katarzyna Koltowska et al.
Development (Cambridge, England), 140(9), 1912-1918 (2013-03-22)
Tightly controlled DNA replication and RNA transcription are essential for differentiation and tissue growth in multicellular organisms. Histone chaperones, including the FACT (facilitates chromatin transcription) complex, are central for these processes and act by mediating DNA access through nucleosome reorganisation.
Cell-cell interactions influence vascular reprogramming by Prox1 during embryonic development.
Kim, H; Cruz, M; Bourdeau, A; Dumont, DJ
Testing null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service