Skip to Content
Merck
All Photos(1)

Key Documents

05-891

Sigma-Aldrich

Anti-phospho-ATF2 (Thr69/71) Antibody, clone AW65

clone AW65, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

AW65, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr69/pThr71)

Gene Information

human ... ATF2(1386)

Specificity

ATF2 phosphorylated at both Thr69 and Thr71
Predicted to cross-react with rat based on sequence homology

Immunogen

KLH-conjugated, synthetic peptide containing the sequence ...DQ[pT]P[pT]P... in which [pT] corresponds to phospho-threonine at residues 69 and 71 of human ATF2

Application

Anti-phospho-ATF2 (Thr69/71) Antibody, clone AW65 is a high quality Mouse Monoclonal Antibody for the detection of phospho-ATF2 (Thr69/71) & has been validated in IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

routinely evaluated by immunoblot with RIPA lysates from anisomycin treated 3T3/NIH cells

Target description

Mr 65kDa

Linkage

Replaces: 04-199

Physical form

Format: Purified
Protein G Purified

Storage and Stability

stable 2 years at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuhui Zheng et al.
Cardiovascular pharmacology: open access, 4(1) (2015-04-01)
The cell type that normally limits the inflammatory and atherosclerotic process is the vascular endothelial cell (EC) that can be regulated by proinflammatory and various stresses. Toll-like receptor-4 (TLR4) plays an important role in the pathogenesis of atherosclerosis, in part
Mumtaz Y Balkhi et al.
iScience, 2, 105-122 (2018-11-15)
T cells infiltrate affected organs in chronic infections and malignancy, but they may fail to eradicate virus-infected cells or tumor because of exhaustion. This report describes a Yin Yang-1 (YY1)-centered mechanism for diverse components that have been correlated with exhaustion.
Ahmad Salameh et al.
The Journal of biological chemistry, 285(30), 23096-23104 (2010-05-29)
Growth factor stimulation induces c-Jun-dependent survival of primary endothelial cells. However, the mechanism of c-Jun anti-apoptotic activity has not been identified. We here demonstrate that in response to growth factor treatment, primary human endothelial cells as well as mouse fibroblasts
Mireille Delhase et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(4), E177-E186 (2011-12-29)
The decision between survival and death in cells exposed to TNF relies on a highly regulated equilibrium between proapoptotic and antiapoptotic factors. The TNF-activated antiapoptotic response depends on several transcription factors, including NF-κB and its RelA/p65 subunit, that are activated
Klára Kirsch et al.
Nature communications, 11(1), 5769-5769 (2020-11-15)
Transcription factor phosphorylation at specific sites often activates gene expression, but how environmental cues quantitatively control transcription is not well-understood. Activating protein 1 transcription factors are phosphorylated by mitogen-activated protein kinases (MAPK) in their transactivation domains (TAD) at so-called phosphoswitches

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service