Documents
Z648442
Quickfit® bends with vacuum connection
socket joint: ST/NS 19/26, cone joint: ST/NS 24/29
Synonym(s):
RA3/23/SC
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Joint
cone joint: ST/NS24/29
socket joint: ST/NS19/26
manufacturer/tradename
Quickfit RA 3/23/SC
availability
not available in USA and Canada
not available in USA and Canada
General description
Socket to cone. With screw-thread adapter for flexible tubing.
Features and Benefits
• Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)• Items marked with * complies with DIN 12594• Designed to fit the angled condenser in vacuum distillation applications• Delivery stem within cone• Supplied with detachable plastic screw thread connectors which allow 9mm bore flexible tubing to be fitted easily and safely
Legal Information
Quickfit is a registered trademark of SciLabware Limited