Key Documents
Z341681
3M 6000/7000 series filters
ABEK1 Organic, ammonia, inorganic and acid gas filter, pkg of 2 ea
About This Item
UNSPSC Code:
46182002
NACRES:
NB.35
Pricing and availability is not currently available.
description
Protects against organic vapours with b.p. >65°C, Ammonia, inorganic and acid gases up to 10 × OEL* or 1000ppm*, EN14387
packaging
pkg of 2 ea
manufacturer/tradename
3M 6059
availability
not available in USA and Canada
General description
Gas and vapour filters
Two filters required per respirator.
Features and Benefits
These filters utilise 3M′s advanced filter technology which minimises breathing resistance for greater comfort to the user. Select gas and vapour filters, particulate filters according to protection required. All filters fit both 6000 and 7000 series respirators by bayonet connection. Two filters required per respirator.
Warning
All these filters should only be used for protection against hazards with good warning properties, e.g. smell or taste, and providing the concentrations of hazardous substances are not immediately dangerous to life or health (IDLH). *Where concentrations are given in terms of the OEL or ppm, the filter should only be used for protection against concentrations below the lower value.
Legal Information
3M is a trademark of 3M Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)