Skip to Content
Merck
All Photos(1)

Key Documents

Z163570

MiniPax® absorbent packets

indicating silica gel, weight 2 g (desiccant), L × W 7/8 in. × 1 3/4 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
24141516
Pricing and availability is not currently available.

manufacturer/tradename

Multisorb 02-00040AG86

L × W

7/8 in. × 1 3/4 in.

weight

2 g (desiccant)

Looking for similar products? Visit Product Comparison Guide

General description

MiniPax® packets are compact, extremely durable, non-dusting, and non-linting with high water vapor trasmission rate and controlled porosity. The packets take up little space in packaging and have a high moisture adsorbing capacity.

Envelopes are formed from sheet of polyethylene fibers has high wet strength and is permeable to both gases and odors. Indicating MiniPax® contain cobalt chloride and should not be used for food or pharmaceutical applications.

  • Compact - The low profile and compact shape of MiniPax® packets saves space and displaces a minimum amount of product. Its flat contour eliminates any potential confusion with a capsule or a pill
  • Maximum surface exposure - The exceptionally narrow seal skirt on only two ends gives MiniPax® packets an unusually large adsorptive surface area for its overall size
  • Integral window - An integral transparent strip permits observation of the contained adsorbent and allows effective use of indicating desiccants to show when moisture has been adsorbed and desiccant should be replaced

Legal Information

MiniPax is a registered trademark of Multisorb Technologies, Inc.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service