Key Documents
Z160466
Fluke® multimeter accessories
Test-lead set
About This Item
UNSPSC Code:
41111700
manufacturer/tradename
Fluke
General description
Kit includes: interchangeable tips; 2 alligator clips, 1retractable hook tip, 2 spade lugs, 2 standard pointed tips; shrouded banana plugs; and carrying pouch.
Legal Information
Fluke is a registered trademark of John Fluke Manufacturing Co., Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)