Skip to Content
Merck
All Photos(1)

Key Documents

T39853

Sigma-Aldrich

Tolylene-2,4-diisocyanate

95%

Synonym(s):

2,4-Diisocyanatotoluene, 4-Methyl-1,3-phenylene diisocyanate, 4-Methyl-m-phenylene diisocyanate, BASF LUPRANATE T80, TDI

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NCO)2
CAS Number:
Molecular Weight:
174.16
Beilstein:
744602
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor density

6 (vs air)

vapor pressure

0.03 mmHg ( 25 °C)

Assay

95%

autoignition temp.

>1148 °F

expl. lim.

9.5 %

impurities

4% 2,6-isomer

refractive index

n20/D 1.568 (lit.)

bp

115-120 °C/10 mmHg (lit.)

mp

20-22 °C (lit.)

density

1.214 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1N=C=O)N=C=O

InChI

1S/C9H6N2O2/c1-7-2-3-8(10-5-12)4-9(7)11-6-13/h2-4H,1H3

InChI key

DVKJHBMWWAPEIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in the preparation of polyurethanes[1] and macrocyclic crown ethers.[2]

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Incl. Phenom. Mol. Recog. Chem., 17, 249-249 (1994)
R S Labow et al.
Biomaterials, 16(1), 51-59 (1995-01-01)
The biostability of polyurethanes was evaluated using a human neutrophil cell culture. The polymers were synthesized with 14C radiolabelled components incorporated into the polyurethane chain and the amount of radiolabel released during exposure to cells and medium was used as
Norman Mangner et al.
JACC. Cardiovascular imaging, 7(12), 1198-1205 (2014-10-13)
The aim of our study was to assess geometric and functional changes of the heart in obese compared with nonobese children and adolescents. Obesity in children and adolescents has increased over the past decades and is considered a strong risk
Tatiana Kuznetsova et al.
American journal of hypertension, 28(4), 535-545 (2014-09-23)
The cardio-renal interaction occurs via hemodynamic and humoral factors. Noninvasive assessment of renal hemodynamics is currently possible by assessment of renal resistive index (RRI) derived from intrarenal Doppler arterial waveforms as ((peak systolic velocity - end-diastolic velocity)/peak systolic velocity). Limited
Anne-Mari Moilanen et al.
PloS one, 10(4), e0124907-e0124907 (2015-04-29)
In a recent genome-wide association study, WD-repeat domain 12 (WDR12) was associated with early-onset myocardial infarction (MI). However, the function of WDR12 in the heart is unknown. We characterized cardiac expression of WDR12, used adenovirus-mediated WDR12 gene delivery to examine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service