D69602
1,5-Dichloropentane
99%
Synonym(s):
Pentamethylene dichloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Cl(CH2)5Cl
CAS Number:
Molecular Weight:
141.04
Beilstein:
1732340
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
99%
form
liquid
refractive index
n20/D 1.457 (lit.)
bp
63-66 °C/10 mmHg (lit.)
mp
−72 °C (lit.)
density
1.106 g/mL at 25 °C (lit.)
SMILES string
ClCCCCCCl
InChI
1S/C5H10Cl2/c6-4-2-1-3-5-7/h1-5H2
InChI key
LBKDGROORAKTLC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
78.8 °F - closed cup
Flash Point(C)
26 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Joshua D Kittle et al.
ACS omega, 5(31), 19820-19826 (2020-08-18)
Selectivity remains a challenge for rapid optical vapor sensing via light reflected from porous silicon photonic crystals. This work highlights a method to increase optical vapor selectivity of porous silicon rugate filters by analyzing additive spectra from two rugate filter
Joshua Kittle et al.
Sensors (Basel, Switzerland), 20(1) (2019-12-29)
Vapor sensing via light reflected from photonic crystals has been increasingly studied as a means to rapidly identify analytes, though few studies have characterized vapor mixtures or chemical warfare agent simulants via this technique. In this work, light reflected from
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service