All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C2H7NO
Molecular Weight:
61.08
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
description
AldrichCPR
form
solid
SMILES string
CCON
InChI
1S/C2H7NO/c1-2-4-3/h2-3H2,1H3
InChI key
AQFWNELGMODZGC-UHFFFAOYSA-N
Other Notes
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
13 - Non Combustible Solids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
José Antonio García et al.
Journal of physiology and biochemistry, 73(2), 235-244 (2017-01-23)
NOS isoform activation is related to liver failure during sepsis, but the mechanisms driving mitochondrial impairment remain unclear. We induced sepsis by LPS administration to inducible nitric oxide synthase (iNOS
Lucie Stuchlíková et al.
Drug testing and analysis, 8(2), 214-220 (2015-04-30)
The present in vitro study was designed to test and compare anthelmintic activity, hepatotoxicity, and biotransformation of four selected aminoacetonitrile derivatives (AADs): monepantel (MOP, anthelmintic approved for the treatment), AAD-970, AAD-1154, and AAD-1336. Micro-agar larval development test, MTT test of
Po-An Yeh et al.
Human molecular genetics, 27(14), 2546-2562 (2018-05-05)
Expansion of poly-glutamine (polyQ) stretches in several proteins has been linked to neurodegenerative diseases. The effects of polyQ-expanded proteins on neurons have been extensively studied, but their effects on glia remain unclear. We found that expression of distinct polyQ proteins
R Ohara et al.
Clinical pharmacology and therapeutics, 101(6), 754-762 (2016-11-20)
Drugs under development can cause unpredicted toxicity in humans due to differential drug responsiveness between humans and other disease models, resulting in clinical trial failures. Human induced pluripotent stem cells (iPSCs) are expected to represent a useful tool for toxicity
Zhihan Zhao et al.
Clinical pharmacology and therapeutics, 106(3), 642-651 (2019-04-05)
Short QT syndrome (SQTS) predisposes afflicted patients to sudden cardiac death. Until now, only one drug-quinidine-has been shown to be effective in patients with SQTS type 1(SQTS1). The objective of this study was to use human-induced pluripotent stem cell-derived cardiomyocytes
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service