Skip to Content
Merck
All Photos(1)

Key Documents

795615

Sigma-Aldrich

Pentamethylcyclopentadienyl cyclopentadienyl rhodium dimer

Synonym(s):

1,1,1,1,1-Pentamethylrhodocene dimer, [RhCp*Cp]2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H40Rh2
Molecular Weight:
606.45
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

Quality Level

storage temp.

−20°C

SMILES string

C[C]1[C](C)[C](C)[C](C)[C]1C.CC2=C(C)C(C)=C(C)C2(C)C3C=CC=C3.[C]4[C][C][C][C]4.[Rh].[Rh]

InChI

1S/C15H20.C10H15.C5H5.2Rh/c1-10-11(2)13(4)15(5,12(10)3)14-8-6-7-9-14;1-6-7(2)9(4)10(5)8(6)3;1-2-4-5-3-1;;/h6-9,14H,1-5H3;1-5H3;1-5H;;

InChI key

WQFYUUJDMAQBRZ-UHFFFAOYSA-N

Application

Used as n-dopant

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production of heavily n- and p-doped CVD graphene with solution-processed redox-active metal?organic species
Paniagua, S. A.;
Materials Horizons, 1, 111-111 (2014)
n-Doping of Organic Electronic Materials Using Air-Stable Organometallics: A Mechanistic Study of Reduction by Dimeric Sandwich Compounds
Guo, S.;
Chemistry (Weinheim An Der Bergstrasse, Germany), 18, 14760-14760 (2012)
Song Guo et al.
Advanced materials (Deerfield Beach, Fla.), 24(5), 699-703 (2011-11-08)
Air-stable dimers of sandwich compounds including rhodocene and (pentamethylcyclopentadienyl)(arene)ruthenium and iron derivatives can be used for n-doping electron-transport materials with electron affinities as small as 2.8 eV. A p-i-n homojunction diode based on copper phthalocyanine and using rhodocene dimer as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service