Skip to Content
Merck
All Photos(1)

Key Documents

771546

Sigma-Aldrich

Sodium bis(trimethylsilyl)amide solution

0.5 M in methyl tert-butyl ether

Synonym(s):

Hexamethyldisilazane sodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3Si]2NNa
CAS Number:
Molecular Weight:
183.37
Beilstein:
3629917
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:

form

liquid

concentration

0.5 M in methyl tert-butyl ether

density

0.761 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C[Si](C)(C)N([Na])[Si](C)(C)C

InChI

1S/C6H18NSi2.Na/c1-8(2,3)7-9(4,5)6;/h1-6H3;/q-1;+1

InChI key

WRIKHQLVHPKCJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Strong base for kinetically controlled deprotonations.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-27.4 °F - closed cup

Flash Point(C)

-33.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniele Fabbri et al.
Environmental science & technology, 42(8), 2957-2963 (2008-05-24)
Using the pyrolysis-gas chromatography-mass spectrometry and off-line pyrolysis/silylation methods for lignites from three Miocene brown coal basins of Poland resulted in the characterization of many organic compounds, including dominant cellulose degradation products such as levoglucosan, 1,6-anhydro-beta-D-glucofuranose, and 1,4:3,6-dianhydroglucopyranose. Levoglucosan is
Hexamethyldisilazane-mediated controlled polymerization of alpha-amino acid N-carboxyanhydrides.
Hua Lu et al.
Journal of the American Chemical Society, 129(46), 14114-14115 (2007-10-30)
A Sebok et al.
Journal of chromatography. A, 1211(1-2), 104-112 (2008-10-14)
This paper presents a derivatization, mass fragmentation study relating to the most common six cholic acids, such as cholic, lithocholic, chenodeoxycholic, ursodeoxycholic, 3-hydroxy,7-ketocholanic and dehydrocholic acids, identified and quantified as pollutants in the aquatic environment at the first time. Derivatizations
Jerry Isaacson et al.
Angewandte Chemie (International ed. in English), 48(10), 1845-1848 (2009-01-29)
(-)-Dysibetaine has been synthesized in 11 steps from readily available L-malic acid (see scheme). The key step is a unique Ugi 4-center-3-component cyclization reaction, where an ester group acts as the carboxylic acid component. The use of 1,1,1,3,3,3-hexamethyldisilazane as an
Natalie M Clark et al.
Chemical communications (Cambridge, England), (39)(39), 5835-5837 (2009-09-30)
'Conventional' (-)-sparteine adducts of lithium and sodium 1,1,1,3,3,3-hexamethyldisilazide (HMDS) were prepared and characterised, along with an unexpected and 'unconventional' hydroxyl-incorporated sodium sodiate, [(-)-sparteine x Na(mu-HMDS)Na x (-)-sparteine](+)[Na(4)(mu-HMDS)(4)(OH)](-)--the complex anion of which is the first inverse crown ether anion.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service