768359
Zirconium(IV) oxide-yttria stabilized
nanopowder, contains 20 mol % yttria as stabilizer, 99.99% trace metals basis
Synonym(s):
20YSZ, YSZ20, YSZ, Yttria stabilized zirconia, Yttrium stabilized zirconium oxide
About This Item
Recommended Products
Assay
99.99% trace metals basis
form
nanopowder
contains
20 mol % yttria as stabilizer
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
greener alternative category
General description
Application
- a thermal barrier coating for the protection and insulation of metal components in gas turbines
- an electrolyte in the fabrication of solid oxide fuel cells (SOFCs)
- in the fabrication of zirconia dental implant abutments
- membranes for micro energy conversion devices
- a solid acid and a catalyst for catalytic applications
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service