Skip to Content
Merck
All Photos(1)

Key Documents

761877

Sigma-Aldrich

4-Methylpyridine-2-carboxaldehyde

97%

Synonym(s):

2-Formyl-4-methylpyridine, 4-Methylpicolinaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO
CAS Number:
Molecular Weight:
121.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

liquid

refractive index

n20/D 1.531

density

1.082 g/mL at 25 °C

functional group

aldehyde

SMILES string

Cc1ccnc(C=O)c1

InChI

1S/C7H7NO/c1-6-2-3-8-7(4-6)5-9/h2-5H,1H3

InChI key

UAKMHSRHDUBNJR-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

186.1 °F

Flash Point(C)

85.6 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service