Skip to Content
Merck
All Photos(1)

Key Documents

741485

Sigma-Aldrich

2-Amino-6-bromo-4-chlorophenol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5BrClNO
CAS Number:
Molecular Weight:
222.47
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

83-87 °C

functional group

bromo
chloro

storage temp.

2-8°C

SMILES string

Nc1cc(Cl)cc(Br)c1O

InChI

1S/C6H5BrClNO/c7-4-1-3(8)2-5(9)6(4)10/h1-2,10H,9H2

InChI key

ISKCLWHHJKNMKI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service