Skip to Content
Merck
All Photos(1)

Key Documents

730289

Sigma-Aldrich

Poly(ethylene glycol) methyl ether acrylate

average Mn 5,000, contains MEHQ as inhibitor

Synonym(s):

Acryl-PEG, Methoxy PEG acrylate, Methoxy poly(ethylene glycol) monoacrylate, Poly(ethylene glycol) monomethyl ether monoacrylate, mPEG-acrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2(CH2CH2O)nCH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

form

solid

mol wt

average Mn 5,000

contains

MEHQ as inhibitor

reaction suitability

reagent type: chemical modification reagent
reaction type: Polymerization Reactions

transition temp

Tm 60-64 °C

density

1.09 g/mL at 25 °C (lit.)

Mw/Mn

<1.1

Ω-end

acrylate

α-end

methoxy

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Synthesized with an initial concentration of ≤5,500 ppm MEHQ

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service