Skip to Content
Merck
All Photos(1)

Key Documents

721913

Supelco

Gadolinium(III) chloride solution

0.1 mg/mL in D2O (99.9 atom % D), water 1 %, Methanol-13C 0.1 % (99 atom % 13C), NMR tube size 5 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GdCl3
CAS Number:
Molecular Weight:
263.61
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.12

composition

Methanol-13C, 0.1% (99 atom % 13C)
water, 1%

concentration

0.1 mg/mL in D2O (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

suitability

suitable for NMR (reference standard)

format

single component solution

SMILES string

Cl[Gd](Cl)Cl

InChI

1S/3ClH.Gd/h3*1H;/q;;;+3/p-3

InChI key

MEANOSLIBWSCIT-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Auto test

Quantity

5 mm O.D. tube contains 0.700 mL.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xianjing Li et al.
Immunologic research, 62(2), 137-145 (2015-04-08)
In this study, we found that pretreatment with low dose of lipopolysaccharide (LPS), also known as lipoglycans and endotoxin, obviously attenuated liver injury caused by diethylnitrosamine (DEN) in mice. This protective effect was described by decreased ALT, TNF-α, and IL-1β
Xiaojuan Chai et al.
Nature communications, 6, 7979-7979 (2015-08-11)
Sepsis is the host's deleterious systemic inflammatory response to microbial infections. Here we report an essential role for the oestrogen sulfotransferase (EST or SULT1E1), a conjugating enzyme that sulfonates and deactivates estrogens, in sepsis response. Both the caecal ligation and
Yan Yang et al.
European journal of immunology, 44(8), 2468-2477 (2014-04-30)
Basophils have been shown to contribute to anaphylaxis through either an IgE-FcεRI-dependent pathway or an IgG-FcγR pathway. However, it remains largely unclear whether basophils can be activated to promote anaphylaxis via a non-FcR pathway as well. The glycolipid receptor ASGM1
Hiroaki Kito et al.
Biochemical and biophysical research communications, 459(3), 457-462 (2015-03-10)
Store-operated Ca(2+) entry (SOCE) via Orai1 and STIM1 complex is supposed to have obligatory roles in the regulation of cellular functions of vascular endothelial cells, while little is known about the contribution of Orai2. Quantitative PCR and Western blot analyses
Quirin Zangl et al.
Anesthesiology, 121(6), 1217-1225 (2014-09-17)
Liver damage by ischemia and reperfusion injury is a risk factor for morbidity and mortality after liver surgery. Postoperative oxygen treatment is routinely applied in the postanesthesia and intensive care unit after liver surgery. The risks of aggravating the injury

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service