Skip to Content
Merck
All Photos(1)

Key Documents

710547

Sigma-Aldrich

Tris(2,4-dimethylphenyl)phosphine

97%

Synonym(s):

Tri-2,4-xylylphosphine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H27P
CAS Number:
Molecular Weight:
346.44
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Assay

97%

reaction suitability

reagent type: ligand

mp

158-162 °C

functional group

phosphine

SMILES string

Cc1ccc(P(c2ccc(C)cc2C)c3ccc(C)cc3C)c(C)c1

InChI

1S/C24H27P/c1-16-7-10-22(19(4)13-16)25(23-11-8-17(2)14-20(23)5)24-12-9-18(3)15-21(24)6/h7-15H,1-6H3

InChI key

XDHRVAHAGMMFMC-UHFFFAOYSA-N

Application

Reactant for sulfonylation reactions

Catalyst for epoxy resin curing

Ligand precatalyst for Heck coupling

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service