Skip to Content
Merck
All Photos(1)

Key Documents

690651

Sigma-Aldrich

(S)-(−)-2-Azido-6-(Boc-amino)hexanoic acid (dicyclohexylammonium) salt

≥97%

Synonym(s):

(S)-2-Azido-6-(Boc-amino)caproic acid (dicyclohexylammonium) salt, N3-Lys(Boc)-OH (dicyclohexylammonium) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20N4O4 · C12H23N
CAS Number:
Molecular Weight:
453.62
Beilstein:
8777290
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%
≥97.0% (HPLC)

form

crystals

optical activity

[α]/D -13.5±1.0°, c = 1% in ethanol

reaction suitability

reaction type: click chemistry

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

C1CCC(CC1)NC2CCCCC2.CC(C)(C)OC(=O)NCCCC[C@H](N=[N+]=[N-])C(O)=O

InChI

1S/C12H23N.C11H20N4O4/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-11(2,3)19-10(18)13-7-5-4-6-8(9(16)17)14-15-12/h11-13H,1-10H2;8H,4-7H2,1-3H3,(H,13,18)(H,16,17)/t;8-/m.0/s1

InChI key

NQXUVISGWVTYSX-WDBKTSHHSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu-Yun Liu et al.
International journal of systematic and evolutionary microbiology, 64(Pt 9), 3247-3253 (2014-07-02)
A methyl parathion (MP) degrading bacterial strain, designated MP-1(T), was isolated from a waste land where pesticides were formerly manufactured in Jiangsu province, China. Polyphasic taxonomic studies showed that MP-1(T) is a Gram-stain-negative, non-spore-forming, rod-shaped and motile bacterium. The bacterium
Marie-Cécile Le Deley et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 32(23), 2440-2448 (2014-07-02)
Relative efficacy and toxicity of cyclophosphamide compared with ifosfamide are debatable. The Euro-EWING99-R1 trial asked whether cyclophosphamide may replace ifosfamide in combination with vincristine and dactinomycin (vincristine, dactinomycin, and cyclophosphamide [VAC] v vincristine, dactinomycin, and ifosfamide [VAI]) after an intensive
Daniel J Henderson et al.
Journal of orthopaedic trauma, 29(2), e54-e59 (2014-07-02)
To analyze functional and radiologic outcomes of tibial shaft fractures treated with the Taylor spatial frame (TSF). Prospective follow-up study of radiologic and functional outcomes. University teaching hospital. From January 2006 to December 2012, prospective data were collected for 56
Tom Bond et al.
Journal of hazardous materials, 278, 288-296 (2014-07-02)
This study investigated the relative effect of chlorination and chloramination on DBP formation from seven model amine precursor compounds, representative of those commonly found in natural waters, at pH 6, 7 and 8. The quantified DBPs included chloroform, dichloroacetonitrile (DCAN)
Catherine Chamberlain et al.
Maternal and child health journal, 19(3), 651-663 (2014-07-02)
Women with gestational diabetes have a high risk of type 2 diabetes postpartum, with Indigenous women particularly affected. This study reports postpartum diabetes screening rates among Indigenous and non-Indigenous women with gestational diabetes, in Far North Queensland, Australia. Retrospective study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service