Skip to Content
Merck
All Photos(1)

Key Documents

686298

Sigma-Aldrich

(R)-4-Chloro-α-methylbenzyl alcohol

95%

Synonym(s):

(R)-1-(4-Chlorophenyl)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH(CH3)OH
CAS Number:
Molecular Weight:
156.61
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

form

liquid

optical activity

[α]20/D +49.0°, c = 1.6 in chloroform

refractive index

n20/D 1.544

density

1.158 g/mL at 25 °C

functional group

chloro
hydroxyl

SMILES string

C[C@@H](O)c1ccc(Cl)cc1

InChI

1S/C8H9ClO/c1-6(10)7-2-4-8(9)5-3-7/h2-6,10H,1H3/t6-/m1/s1

InChI key

MVOSNPUNXINWAD-ZCFIWIBFSA-N

Application

(R)-4-Chloro-α-methylbenzyl alcohol can be used:
  • As a substrate in the racemization process of secondary alcohols using ruthenium hydroxide complexes.[1]
  • In the synthesis of 2,3,4,9-tetrahydro-1H-carbazole as potent DP1 antagonists.[2]
  • As a substrate in the synthesis of chiral 3-aryl-3-substituted propanoic acids through Mitsunobu reaction.[3]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective Carbon- Carbon Bond Formation via the Mitsunobu Displacement of Chiral Secondary Benzylic Alcohols
Hillier MC, et al.
Organic Letters, 6(4), 573-576 (2004)
Ruthenium Hydroxide Complexes in the Racemization of Secondary Alcohols
Nun P, et al.
Organometallics, 30(23), 6347-6350 (2011)
Potent and highly selective DP1 antagonists with 2, 3, 4, 9-tetrahydro-1H-carbazole as pharmacophore
Li L, et al.
Bioorganic & Medicinal Chemistry Letters, 20(24), 7462-7465 (2010)
Xiao-Hong Chen et al.
Microbial cell factories, 11, 119-119 (2012-09-06)
The bacterium Acetobacter sp. CCTCC M209061 is a promising whole-cell biocatalyst with exclusive anti-Prelog stereoselectivity for the reduction of prochiral ketones that can be used to make valuable chiral alcohols such as (R)-4-(trimethylsilyl)-3-butyn-2-ol. Although it has promising catalytic properties, its
Xiao-Hong Chen et al.
BMC biotechnology, 11, 110-110 (2011-11-22)
Chiral alcohols are widely used in the synthesis of chiral pharmaceuticals, flavors and functional materials and appropriate whole-cell biocatalysts offer a highly enantioselective, minimally polluting route to these valuable compounds. The recently isolated strain Acetobacter sp. CCTCC M209061 showed exclusive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service