Skip to Content
Merck
All Photos(1)

Key Documents

548758

Sigma-Aldrich

Methyl thiazolidine-2-carboxylate hydrochloride

98%

Synonym(s):

Thiazolidine-2-carboxylic acid methyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2S · HCl
CAS Number:
Molecular Weight:
183.66
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

159-163 °C (dec.) (lit.)

functional group

ester
thioether

SMILES string

Cl[H].COC(=O)C1NCCS1

InChI

1S/C5H9NO2S.ClH/c1-8-5(7)4-6-2-3-9-4;/h4,6H,2-3H2,1H3;1H

InChI key

YGTQIXUXZVNVKG-UHFFFAOYSA-N

General description

Thiazolidine-2-carboxylic acid methyl ester hydrochloride is a cyclic secondary amino acid derivative. The thiazolidine-2-carboxylic acid (Thz) moiety can exhibit ACE (angiotensin converting enzyme) inhibitor activity in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C J Blankley et al.
Journal of medicinal chemistry, 30(6), 992-998 (1987-06-01)
The synthesis of a series of novel, potent angiotensin converting enzyme (ACE) inhibitors containing saturated bicyclic amino acids in place of proline is described. Octahydroindole-2-carboxylic acid, octahydroisoindole-1-carboxylic acid, and octahydro-3-oxoisoindole-1-carboxylic acid can replace proline in both sulfhydryl and non-sulfhydryl ACE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service