Skip to Content
Merck
All Photos(2)

Key Documents

547816

Sigma-Aldrich

Tris[N,N-bis(trimethylsilyl)amide]europium(III)

98%

Synonym(s):

1,1,1-Trimethyl-N-(trimethylsilyl)silanamine europium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Eu(N(Si(CH3)3)2)3
CAS Number:
Molecular Weight:
633.12
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

reaction suitability

core: europium
reagent type: catalyst

mp

147-154 °C (lit.)

SMILES string

C[Si](C)(C)N([Eu](N([Si](C)(C)C)[Si](C)(C)C)N([Si](C)(C)C)[Si](C)(C)C)[Si](C)(C)C

InChI

1S/3C6H18NSi2.Eu/c3*1-8(2,3)7-9(4,5)6;/h3*1-6H3;/q3*-1;+3

InChI key

FLADHQMJJLOUHM-UHFFFAOYSA-N

Application

Reactant for:
  • Preferential complexation reactions
  • Pin-wheel shaped heterobimetallis lanthanide alkali metal binaphtholates

Reactant for synthesis of:
  • Isostructural neo-pentoxide derivatives of Group 3 and lanthanide series metals for production of Ln2O3 nanoparticles
  • Anhydrous mononuclear tris(8-quinolinolato)lanthanoid complexes for organic light-emitting devices

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

36.0 °F - closed cup

Flash Point(C)

2.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elsa Vennat et al.
Dental materials : official publication of the Academy of Dental Materials, 25(6), 729-735 (2009-01-29)
The objectives of this study were to assess demineralized dentin porosity and quantify the different porous features distribution within the material using mercury intrusion porosimetry (MIP) technique. We compared hexamethyldisilazane (HMDS) drying and lyophilization (LYO) (freeze-drying) in sample preparation. Fifty-six
Nur Hazlin Hazrin-Chong et al.
Journal of microbiological methods, 90(2), 96-99 (2012-05-09)
The use of hexamethyldisilazane (HMDS) as a drying agent was investigated in the specimen preparation for scanning electron microscopy (SEM) imaging of bacterial surface colonization on sub-bituminous coal. The ability of microbes to biofragment, ferment and generate methane from coal
Nan Li et al.
Lab on a chip, 8(12), 2105-2112 (2008-11-22)
High-density live cell array serves as a valuable tool for the development of high-throughput immunophenotyping systems and cell-based biosensors. In this paper, we have, for the first time, demonstrated a simple fabrication process to form the hexamethyldisilazane (HMDS) and poly(ethylene
Hung-Che Chou et al.
Colloids and surfaces. B, Biointerfaces, 73(2), 244-249 (2009-06-24)
This paper utilizes a silver electrode quartz crystal microbalance (QCM) mass sensor to detect the physiology of cells. This study also investigates the plasma surface modification of silver electrode QCMs through deposition of hexamethyldisilazane (HMDSZ) films as a protection film.
S V Buravkov et al.
Bulletin of experimental biology and medicine, 151(3), 378-382 (2012-03-28)
We compared conventional method for specimen (cell cultures, tissue specimens) preparation for scanning electron microscopy and a method without sputtering and critical-point drying. OTO-method (osmium-thiocarbohydrazide-osmium) with sample impregnation with hexamethyldisilazane followed by air drying was used as an alternative method.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service