Skip to Content
Merck
All Photos(1)

Key Documents

476021

Sigma-Aldrich

2-Methoxyphenethylamine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2CH2NH2
CAS Number:
Molecular Weight:
151.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.541

bp

236-237 °C (lit.)

density

1.033 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

COc1ccccc1CCN

InChI

1S/C9H13NO/c1-11-9-5-3-2-4-8(9)6-7-10/h2-5H,6-7,10H2,1H3

InChI key

WSWPCNMLEVZGSM-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

2-Methoxyphenethylamine (MPEA) is also referred to as o-methoxyphenethylamine. The distribution behavior of tyramine and MPEA in the [bmim][PF6]/water system (bmim = 1-butyl-3-methylimidazolium hexafluorophosphate) has been evaluated under different conditions.[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave-enhanced cyanation of aryl halides with a dimeric ortho-palladated complex catalyst.
Hajipour AR, et al.
Transition Met. Chem. (London), 36(7), 725-730 (2011)
The Partitioning Behavior of Tyramine and 2-Methoxyphenethylamine in a Room Temperature Ionic Liquid-Water System Compared to Traditional Organic-Water System.
Yung KL, et al.
Separation Science and Technology, 40(7), 1555-1566 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service