Skip to Content
Merck
All Photos(1)

Key Documents

451282

Sigma-Aldrich

Strontium chloride

AnhydroBeads, −10 mesh, 99.995% trace metals basis

Synonym(s):

Strontium dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrCl2
CAS Number:
Molecular Weight:
158.53
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for synthesis
synthesis grade

grade

for synthesis
synthesis grade

product line

AnhydroBeads

Assay

99.995% trace metals basis

impurities

≤55.0 ppm Trace Metal Analysis

particle size

−10 mesh

bp

1250 °C/1 atm (lit.)

mp

874 °C (lit.)

density

3 g/mL at 25 °C (lit.)

SMILES string

Cl[Sr]Cl

InChI

1S/2ClH.Sr/h2*1H;/q;;+2/p-2

InChI key

AHBGXTDRMVNFER-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

T C Brennan et al.
British journal of pharmacology, 157(7), 1291-1300 (2009-07-01)
Strontium ranelate reduces fracture risk in postmenopausal women with osteoporosis. Evidence from non-clinical studies and analyses of bone markers in phase III trials indicate that this is due to an increase in osteoblast formation and a decrease of osteoclastic resorption.
Jing Chen et al.
PLoS biology, 19(4), e3001134-e3001134 (2021-04-27)
Cell death is a vital event in life. Infections and injuries cause lytic cell death, which gives rise to danger signals that can further induce cell death, inflammation, and tissue damage. The mevalonate (MVA) pathway is an essential, highly conserved
Tao Hu et al.
Spine, 40(9), 613-621 (2015-02-24)
A rodent posterolateral spinal fusion model. This study evaluated a protamine-based polyelectrolyte complex (PEC) developed to use heparin in enhancing the biological activity of low-dose recombinant human bone morphogenetic protein-2 (rhBMP-2) in spinal fusion. rhBMP-2 is commonly regarded as the
Fan Yang et al.
Stem cells (Dayton, Ohio), 29(6), 981-991 (2011-05-13)
Strontium ranelate is a newly approved drug that can reduce the risk of vertebral fracture, which is attributed to its dual function in increasing the bone formation and decreasing the bone resorption. Strontium-containing hydroxyapatite was also demonstrated to stimulate the
Karen Versieren et al.
Reproductive biomedicine online, 21(6), 769-775 (2010-11-06)
Parthenogenetic reconstruction is one major strategy to create patient-specific stem cells. The aim of this study was to find the best artificial activation protocol for parthenogenetic activation of mouse and human oocytes comparing different methods. In a first set of

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service