Recommended Products
Assay
95%
form
liquid
refractive index
n20/D 1.511 (lit.)
bp
115 °C/0.015 mmHg (lit.)
density
1.225 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)c1ccc(CCl)o1
InChI
1S/C8H9ClO3/c1-2-11-8(10)7-4-3-6(5-9)12-7/h3-4H,2,5H2,1H3
InChI key
JBACYJRMCXLIQU-UHFFFAOYSA-N
Application
Ethyl 5-(chloromethyl)-2-furancarboxylate may be employed for the synthesis of a series of novel sulfamide- and urea-based small-molecule antagonists of the protein-protein interaction IL-2/IL-2Rα. It may be employed as starting material for the synthesis of 5-hydroxymethylfuran-2-carboxylic acid.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Bioorganic & medicinal chemistry letters, 15(4), 983-987 (2005-02-03)
The identification, design, and synthesis of a series of novel sulfamide- and urea-based small-molecule antagonists of the protein-protein interaction IL-2/IL-2Ralpha are described. Installation of a furan carboxylic acid fragment onto a low-micromolar sulfamide resulted in a 23-fold improvement in activity
PloS one, 7(7), e41142-e41142 (2012-08-01)
Many investigators have recognised that a significant proportion of environmental bacteria exist in a viable but non-culturable state on agar plates, and some researchers have also noticed that some of such bacteria clearly recover their growth on matrices other than
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service