Skip to Content
Merck
All Photos(1)

Key Documents

423327

Sigma-Aldrich

2,4-Di-tert-butyl-6-(5-chloro-2H-benzotriazol-2-yl)phenol

98%

Synonym(s):

2,4-Di-(tert -butyl)-6-(5-chloro-2H -benzo[d ][1,2,3]triazol-2-yl)phenol, 2-(2-Hydroxy-3,5-di-tert -butylphenyl)-5-chloro-2H -benzotriazole, 2-(2-Hydroxy-3,5-di-tert -butylphenyl)-5-chlorobenzotriazole, 2-(5-Chloro-2H -benzotriazol-2-yl)-4,6-bis(1,1-dimethylethyl)phenol, 5-Chloro-2-(3,5-di-tert -butyl-2-hydroxyphenyl)benzotriazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24ClN3O
CAS Number:
Molecular Weight:
357.88
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

mp

150-153 °C (lit.)

SMILES string

CC(C)(C)c1cc(c(O)c(c1)C(C)(C)C)-n2nc3ccc(Cl)cc3n2

InChI

1S/C20H24ClN3O/c1-19(2,3)12-9-14(20(4,5)6)18(25)17(10-12)24-22-15-8-7-13(21)11-16(15)23-24/h7-11,25H,1-6H3

InChI key

UWSMKYBKUPAEJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Di-tert-butyl-6-(5-chloro-2H-benzotriazol-2-yl)phenol belongs to the class of benzotriazole based UV absorbers and UV filters, which can be used in cosmetics and personal care products, such as shampoos, body lotions and sunscreens.

Application

UV absorber

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of UV filters in human breast milk using turbulent flow chromatography and babies? daily intake estimation.
Molins-Delgado D, et al.
Environmental Research, 161, 532-539 (2018)
Multiresidue analytical method for the determination of antimicrobials, preservatives, benzotriazole UV stabilizers, flame retardants and plasticizers in fish using ultra high performance liquid chromatography coupled with tandem mass spectrometry.
Kim J, et al.
Journal of Chromatography A, 1218(22), 3511-3520 (2011)
Zhe Lu et al.
Journal of chromatography. A, 1461, 51-58 (2016-07-19)
Analytical methods were developed for the determination of eight substituted diphenylamines (SDPAs) and six benzotriazole UV stabilizers (BZT-UVs) in blood plasma and fish homogenate matrices. Liquid-liquid extraction by methyl tert-butyl ether and denaturation by KOH following silica gel packed column

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service