Skip to Content
Merck
All Photos(2)

Documents

405701

Sigma-Aldrich

Allyl P,P-diethylphosphonoacetate

97%

Synonym(s):

Phosphonoacetic acid P,P-diethyl allyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2P(O)CH2COOCH2CH=CH2
CAS Number:
Molecular Weight:
236.20
Beilstein:
1788582
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.445 (lit.)

bp

157-158 °C/10 mmHg (lit.)

density

1.12 g/mL at 25 °C (lit.)

SMILES string

CCOP(=O)(CC(=O)OCC=C)OCC

InChI

1S/C9H17O5P/c1-4-7-12-9(10)8-15(11,13-5-2)14-6-3/h4H,1,5-8H2,2-3H3

InChI key

USZLAMOPIWNLPH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Horner-Wadsworth-Emmons reaction
  • Preparation of phosphonate-functionalized cyclopentenones via regioselective Pauson-Khand processes
  • Preparation of salicylihalamide analog saliphenylhalamide as potential vacuolar ATPase inhibitors and anticancer agents
  • Solid-phase synthesis of olefin-containing peptides via Horner-Emmons reaction as inhibitors of HTLV-1 protease
  • Synthesis of fluorescent poly(aromatic amide) dendrimers

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service