Skip to Content
Merck
All Photos(2)

Key Documents

387924

Sigma-Aldrich

Poly(vinyl acetate)

average Mw ~140,000 by GPC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(O2CCH3)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

beads

mol wt

average Mw ~140,000 by GPC

storage temp.

2-8°C

SMILES string

COC(=O)C=C

InChI

1S/C4H6O2/c1-3-6-4(2)5/h3H,1H2,2H3

InChI key

XTXRWKRVRITETP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seon Tae Kim et al.
International journal of pediatric otorhinolaryngology, 77(1), 113-116 (2012-11-08)
The purpose of this prospective study was to determine the effectiveness of polyurethane foam (PUF) and polyvinyl acetate (PA) as packing materials for reducing post-conchotomy bleeding, pain, and headaches. This study was a prospective, randomized and single-blinded controlled study. Fifty-two
Priscileila Colerato Ferrari et al.
Carbohydrate polymers, 91(1), 244-252 (2012-10-10)
In this work pellets containing chitosan for colonic drug delivery were developed. The influence of the polysaccharide in the pellets was evaluated by swelling, drug dissolution and intestinal permeation studies. Drug-loaded pellets containing chitosan as swellable polymer were coated with
Dae Woo Kim et al.
American journal of rhinology & allergy, 26(5), e147-e149 (2012-11-22)
This study investigated the efficacy of glove finger-coated polyvinyl acetate (PA) pack on hemostasis, pain levels, and wound healing after endoscopic sinus surgery (ESS). A prospective, randomized, double-blinded controlled study was performed in 30 patients who underwent bilateral ESS for
Congming Xiao et al.
International journal of biological macromolecules, 52, 349-352 (2012-10-31)
Tailor-made conjunct of methyl cellulose (MC) and polyvinyl acetate (PVAc) was synthesized through the combination of reversible addition-fragmentation chain transfer (RAFT) polymerization and thiol-ene click reaction. MC was firstly transferred into unsaturated MC (UMC), and then covalently connected with well-defined
V Andonova et al.
Die Pharmazie, 67(7), 601-604 (2012-08-15)
During the last decade the number of investigations on the preparation and application of more effective drug release systems on the basis of nanocarriers from biocompatible and biodegradable polymers are considerably increasing. This is notably in force for practically water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service