Skip to Content
Merck
All Photos(1)

Documents

374148

Sigma-Aldrich

N-tert-Butylisopropylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CNHCH(CH3)2
CAS Number:
Molecular Weight:
115.22
Beilstein:
1846752
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.398 (lit.)

bp

98 °C (lit.)

density

0.727 g/mL at 25 °C (lit.)

SMILES string

CC(C)NC(C)(C)C

InChI

1S/C7H17N/c1-6(2)8-7(3,4)5/h6,8H,1-5H3

InChI key

ZWXQPERWRDHCMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-tert-Butylisopropylamine participates in the synthesis of hindered enamines.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David M Hodgson et al.
Journal of the American Chemical Society, 126(22), 6870-6871 (2004-06-04)
A new reactivity mode of lithium amides with epoxides leads to hindered enamines. The reaction of some of these enamines with unactivated primary and secondary alkyl halides is described, which expands the range of electrophiles that one can use in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service