Skip to Content
Merck
All Photos(1)

Key Documents

362557

Sigma-Aldrich

Alanine anhydride

99%

Synonym(s):

3,6-Dimethyl-2,5-piperazinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N2O2
CAS Number:
Molecular Weight:
142.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

description

mixture of DL and meso

Assay

99%

mp

283-285 °C (lit.)

solubility

1 M HCl: soluble 25 mg/mL, clear, colorless to light yellow

SMILES string

CC1NC(=O)C(C)NC1=O

InChI

1S/C6H10N2O2/c1-3-5(9)8-4(2)6(10)7-3/h3-4H,1-2H3,(H,7,10)(H,8,9)

InChI key

WWISPHBAYBECQZ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The synthesis of peptides by transamination.
Herbst RM and Shemin D.
The Journal of Biological Chemistry, 147(3), 541-547 (1943)
Synthesis of Aliphatic Poly (ester-amide) s by Ring-Opening Polymerization of L-Lactide and Amino Acid Anhydrides.
Noomhorma C and Tokiwab Y.
ScienceAsia, 34(1), 43-47 (2008)
Elahe Alizadeh et al.
The Journal of chemical physics, 134(5), 054305-054305 (2011-02-10)
Dissociative electron attachment to dialanine and alanine anhydride has been studied in the gas phase utilizing a double focusing two sector field mass spectrometer. We show that low-energy electrons (i.e., electrons with kinetic energies from near zero up to 13

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service