Skip to Content
Merck
All Photos(1)

Key Documents

339172

Sigma-Aldrich

Fluoroantimonic acid hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSbF6 · 6H2O
CAS Number:
Molecular Weight:
344.85
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid
Pricing and availability is not currently available.

grade

for analytical purposes

vapor pressure

8 mmHg ( 47 °C)

form

solid

concentration

32-34% (fluorine (F))

SMILES string

O.O.O.O.O.O.F.F[Sb](F)(F)(F)F

InChI

1S/6FH.6H2O.Sb/h6*1H;6*1H2;/q;;;;;;;;;;;;+5/p-5

InChI key

WWPSSNXRZMXGHE-UHFFFAOYSA-I

Application

HF/SbF5 is an excellent medium for the preparation of alkylidene oxonium salts.[1]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rolf Minkwitz et al.
Inorganic chemistry, 38(5), 844-847 (2001-10-24)
We have studied the preparation and isolation of the halogenated alkylidene oxonium salts CH(3)OCHX(+)MF(6)(-) (X = Cl, F; M = As, Sb). Addition of dichloromethylmethylether at -78 degrees C to the superacidic media HF/MF(5) resulted in the formation of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service