Skip to Content
Merck
All Photos(1)

Key Documents

301493

Sigma-Aldrich

2,4-Dimethylaniline

98%

Synonym(s):

1-Amino-2,4-dimethylbenzene, 2,4-Xylidine, 4-Amino-m-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
636243
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

4.2 (vs air)

vapor pressure

0.16 mmHg ( 25 °C)

Assay

98%

form

liquid

autoignition temp.

860 °F

expl. lim.

7 %

refractive index

n20/D 1.558 (lit.)

bp

218 °C (lit.)

mp

−14.3 °C (lit.)

density

0.98 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(N)c(C)c1

InChI

1S/C8H11N/c1-6-3-4-8(9)7(2)5-6/h3-5H,9H2,1-2H3

InChI key

CZZZABOKJQXEBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Dimethylaniline is hydrolysis product of amitraz (sum) in pears.[1] Electrochemical method for the determination of total amitraz residues and its final breakdown product, 2,4-dimethylaniline was reported.[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia Hepperle et al.
Food chemistry, 166, 240-247 (2014-07-24)
We developed a method for the hydrolysis of amitraz, and its degradation products 2,4-dimethylphenyl-N'-methylformamidine (DMPF) and 2,4-dimethylformamide (DMF) into 2,4-dimethylaniline (DMA), directly from QuEChERS extracts. The hydrolysis product DMA was analysed using liquid chromatography-electrospray tandem mass spectrometry (LC-ESI-MS/MS) in the
R Brimecombe et al.
Talanta, 71(3), 1298-1303 (2007-02-28)
Amitraz is a formamide acaracide used in the control of ticks and mites in livestock. An electrochemical method for the determination of total amitraz residues and its final breakdown product, 2,4-dimethylaniline, is presented. Cyclic voltammetry at a glassy carbon electrode
Stefan H Bossmann et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 3(5), 401-407 (2002-12-06)
TiO2-codoped [FeII(bpy)3]2+/zeolite Y systems are revealed to be efficient photocatalysts for photochemically enhanced Fenton processes. Preparation, characterization, and tests using 2,4-xylidine as a model pollutant are described, and a first mechanistic hypothesis explaining the rather high photochemical reactivity of the
Ana M Amat et al.
Chemosphere, 57(9), 1123-1130 (2004-10-27)
The possibility of using zeolites containing the 2,4,6-triphenylpyrylium cation as photocatalysts for the degradation of pollutants has been tested on aqueous xylidine (2,4-dimethylaniline) solutions as models for contaminated wastewaters. The influence of the photocatalyst and substrate concentrations on xylidine oxidation
S Göb et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 44(5), 339-345 (2001-11-07)
Among advanced oxidation processes (AOPs), the photochemically enhanced Fenton reaction may be considered as one of the most efficient for the degradation of contaminants in industrial wastewater. This process involves a series of complex reactions. Therefore, an empirical model based

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service