Skip to Content
Merck
All Photos(2)

Key Documents

205400

Sigma-Aldrich

Polychloroprene

85% trans, 10% cis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CH=C(Cl)CH2-]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

chunks

viscosity

100, Mooney(lit.)

transition temp

Tg −48 °C

density

1.23 g/mL at 25 °C (lit.)

SMILES string

ClC(=C)C=C

InChI

1S/C4H5Cl/c1-3-4(2)5/h3H,1-2H2

InChI key

YACLQRRMGMJLJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexis G Davis et al.
Journal of athletic training, 46(6), 615-620 (2011-01-01)
Knee braces and neoprene sleeves are commonly worn by people with anterior cruciate ligament reconstructions (ACLRs) during athletic activity. How knee braces and sleeves affect muscle activation in people with ACLRs is unclear. To determine the effects of knee braces
Anne B-H Dall et al.
Contact dermatitis, 67(2), 89-93 (2012-03-21)
Diethylthiourea is widely used in the rubber industry, particularly in neoprene rubber, and may cause allergic contact dermatitis. However, as thiourea allergens are not part of the European baseline series, the diagnosis of allergic contact dermatitis caused by thiourea compounds
Jin-Rong Zhang et al.
Chemistry & biodiversity, 9(10), 2218-2224 (2012-10-20)
Nine achiral tetraprenylated alkaloids, including three new compounds, named malonganenones I-K (1-3, resp.), together with six known analogs, 4-9, were isolated from the gorgonian Euplexaura robusta collected from Weizhou Island of Guangxi Province, China. The structures of compounds 1-3 were
A linear allergic contact dermatitis to p-tert-butylphenol formaldehyde resin sectorially present in a neoprene orthopedic brace: role of spectroscopy.
Monica Corazza et al.
Dermatitis : contact, atopic, occupational, drug, 23(6), 292-293 (2012-11-22)
Hans Wilhelm Pau et al.
Acta oto-laryngologica, 131(12), 1279-1285 (2011-09-06)
The findings suggest that underwater sound perception is realized by the middle ear rather than by bone conduction, at least in shallow water conditions. To prove whether underwater sound perception is effected by bone conduction or by conduction via the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service