191019
Poly(dimer acid-co-alkyl polyamine)
softening point 95 °C (ring and ball)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
density
0.97 g/mL at 25 °C
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service