About This Item
Recommended Products
Assay
99%
form
solid
mp
68-71 °C (lit.)
functional group
hydroxyl
SMILES string
OC1Cc2ccccc2C1
InChI
1S/C9H10O/c10-9-5-7-3-1-2-4-8(7)6-9/h1-4,9-10H,5-6H2
InChI key
KMGCKSAIIHOKCX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service