Chlordimeform
Synonym(s):
N-(4-Chloro-2-methylphenyl)-N′,N′-dimethylformamidine, Chlorodimeform, Chlorophenamidine
Empirical Formula (Hill Notation):
C10H13ClN2
CAS Number:
Molecular Weight:
196.68
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing