Skip to Content
Merck
All Photos(1)

Key Documents

29162

Sigma-Aldrich

Methyl 2-oxocyclohexanecarboxylate

technical, ≥90% (GC)

Synonym(s):

2-Methoxycarbonylcyclohexanone, Methyl cyclohexanone-2-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O3
CAS Number:
Molecular Weight:
156.18
Beilstein:
510258
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥90% (GC)

refractive index

n20/D 1.483

bp

50 °C/0.2 mmHg (lit.)

density

1.10 g/mL at 20 °C (lit.)

SMILES string

COC(=O)C1CCCCC1=O

InChI

1S/C8H12O3/c1-11-8(10)6-4-2-3-5-7(6)9/h6H,2-5H2,1H3

InChI key

JEENWEAPRWGXSG-UHFFFAOYSA-N

Application

Methyl 2-oxocyclohexanecarboxylate has been used in the synthesis of substituted tetrahydrobenzofuran derivatives via reaction with propargylic esters in the presence of palladium catalyst.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly diastereoselective synthesis of tetrahydrobenzofuran derivatives by palladium-catalyzed reaction of propargylic esters with substituted ?-dicarbonyl compounds.
Yoshida M, et al.
Tetrahedron, 66(14), 2675-2682 (2010)
Qinglei Meng et al.
Nature communications, 8, 14190-14190 (2017-02-01)
Cyclohexanone and its derivatives are very important chemicals, which are currently produced mainly by oxidation of cyclohexane or alkylcyclohexane, hydrogenation of phenols, and alkylation of cyclohexanone. Here we report that bromide salt-modified Pd/C in H

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service