Merck
  • Bone resorption is regulated by cell-autonomous negative feedback loop of Stat5-Dusp axis in the osteoclast.
Bone resorption is regulated by cell-autonomous negative feedback loop of Stat5-Dusp axis in the osteoclast.
The Journal of experimental medicine (2013-12-25)
Jun Hirose, Hironari Masuda, Naoto Tokuyama, Yasunori Omata, Takumi Matsumoto, Tetsuro Yasui, Yuho Kadono, Lothar Hennighausen, Sakae Tanaka
ABSTRACT
Signal transducer and activator of transcription 5 (Stat5) is essential for cytokine-regulated processes such as proliferation, differentiation, and survival in hematopoietic cells. To investigate the role of Stat5 in osteoclasts, we generated mice with an osteoclast-specific conditional deletion of Stat5 (Stat5 conditional knockout [cKO] mice) and analyzed their bone phenotype. Stat5 cKO mice exhibited osteoporosis caused by an increased bone-resorbing activity of osteoclasts. The activity of mitogen-activated protein kinases (MAPKs), in particular extracellular signal-related kinase, was increased in Stat5 cKO osteoclasts, whereas the expression of the MAPK phosphatases dual specificity phosphatase 1 (Dusp1) and Dusp2 was significantly decreased. Interleukin-3 (IL-3) stimulated the phosphorylation and nuclear translocation of Stat5 in osteoclasts, and Stat5 expression was up-regulated in response to receptor activator of nuclear factor κB ligand (RANKL). The results suggest that Stat5 negatively regulates the bone-resorbing function of osteoclasts by promoting Dusp1 and Dusp2 expression, and IL-3 promotes Stat5 activation in osteoclasts.
Sigma-Aldrich
Penicillinase from Bacillus cereus, lyophilized, powder, white, ~13 U/mg
Sigma-Aldrich
β-Lactamase from Enterobacter cloacae, Type IV, lyophilized powder, 0.2-0.6 units/mg protein (using benzylpenicillin)
Sigma-Aldrich
Penicillinase from Bacillus cereus, lyophilized powder, 1,500-3,000 units/mg protein (using benzylpenicillin)
Sigma-Aldrich
β-Lactamase from Pseudomonas fluorescens, lyophilized powder, recombinant, expressed in E. coli
Sigma-Aldrich
β-Lactamase Blend, recombinant, expressed in E. coli
Sigma-Aldrich
β-Lactamase, recombinant, expressed in E. coli