VIV105001
SynVivo SynRAM 3D Inflammation model chip, SMN2 microvascular network configuration
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
60104506
NACRES:
NA.84
Recommended Products
packaging
case of 1 ea
manufacturer/tradename
SynVivo 105001
storage temp.
room temp
General description
SynRAm inflammation model co-culture chip- Microvascular design Barrier type: Pillars, Distance between pillars: 50 μm, Pillar Diameter: 10 μm, Barrier height: 3 μm, Travel/barrier width: 50 μm, Device depth: 100 μm. The SynRAM 3D Inflammation Model has been developed to study the entire inflammation pathway in a realistic and dynamic environment.
Linkage
LInk to Publications
application text
application text
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service