Z549878
Aldrich® jacketed three-neck flask
full jacket, capacity 500 mL, center joint: ST/NS 24/40
Synonym(s):
boiling flask, flask round, flask round bottom
All Photos(2)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
borosilicate glass
round bottom
three-neck
three-neck flask
Joint
center joint: ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
500 mL
Related Categories
General description
Unique design which incorporates a flush-closing PTFE drain valve eliminating unwanted dead space.
Features:
Features:
- Eliminates dead space for complete stirring of flask contents
- Allows for complete draining without breaking down flask set-up
- Liquid cooled or heated with liquid or steam
- 24/40 side joints
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC