Z302899
Quickfit® round-bottom flasks
capacity 500 mL, joint: ST/NS 34/35
Synonym(s):
FR500/5S
All Photos(1)
About This Item
UNSPSC Code:
41121804
Recommended Products
material
borosilicate glass
round bottom flask
short
Joint
joint: ST/NS34/35
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
case of 5
manufacturer/tradename
Quickfit FR500/5S
availability
not available in USA and Canada
capacity
500 mL
General description
Short neck with ground-glass joint.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)