Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300331

Sigma-Aldrich

Anti-STAT6 (Ab-641) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-D12S1644 antibody produced in rabbit, Anti-IL-4-STAT antibody produced in rabbit, Anti-STAT6B antibody produced in rabbit, Anti-STAT6C antibody produced in rabbit, Anti-signal transducer and activator of transcription 6, interleukin-4 induced antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~110 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(R-G-Y-V-P)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STAT6(6778)

Immunogen

Peptide sequence around aa. 639-643 (R-G-Y-V-P), according to the protein STAT6.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

STAT6 is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor-associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. By screening an embryonic lung fibroblast cDNA library with a wildtype STAT6 probe, 2 variant cDNAs were identified, which were termed STAT6B and STAT6C, encoding an N-terminal 110-amino acid truncation and a 27-amino acid deletion in the SH2 domain, respectively. All the three variants are ubiquitously expressed with STAT6B expression greatest in spleen and STAT6C expression greatest in lung. STAT6B resembles an attenuated STAT6, but that STAT6C inhibits IL-4-mediated mitogenesis and cell surface antigen expression, and is not tyrosine phosphorylated.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boon Leong Quah et al.
Virchows Archiv : an international journal of pathology, 478(5), 977-984 (2020-09-13)
Primary non-neuroendocrine tumours of the pituitary gland and sella are rare lesions often challenging to diagnose. We describe two cases of clinically aggressive primary glomus tumour of the pituitary gland. The lesions occurred in a 63-year-old male and a 30-year-old

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service