Skip to Content
Merck
All Photos(1)

Documents

4.90205

Sigma-Aldrich

1-Ethyl-3-methylimidazolium octylsulfate

for synthesis

Synonym(s):

1-Ethyl-3-methylimidazolium octylsulfate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
320.45
UNSPSC Code:
12352107

form

liquid

pH

2.9 ( in H2O)

mp

-9 °C

density

1.095 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H18O4S.C6H11N2/c1-2-3-4-5-6-7-8-12-13(9,10)11;1-3-8-5-4-7(2)6-8/h2-8H2,1H3,(H,9,10,11);4-6H,3H2,1-2H3/q;+1/p-1

InChI key

DAFDIILBFQKIIA-UHFFFAOYSA-M

General description

for synthesis

Analysis Note

Purity (HPLC)≥ 98.0 %Identity (NMR)passes testHalides (IC)≤ 0.1 %Water (K. F.)≤ 1.0 %

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service