Skip to Content
Merck
All Photos(1)

Documents

772690

Sigma-Aldrich

IsoYeast - Growth Medium (Unlabeled)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.12

form

powder

Quality Level

technique(s)

bio NMR: suitable

storage temp.

2-8°C

Application

Each IsoYeast product is a complete system used to prepare dual media - 1 liter of Growth Medium and 250 mL of Expression Medium. The dual media are offered as lyophilized powdered components and need addition of only sterile water to the Growth Medium, and water plus methanol to the Expression Medium.
Stable isotope enrichment of proteins is necessary for study by NMR spectroscopy. One way this can be accomplished is using a methylotrophic yeast (Pichia pastoris) expression system for proteins. The use of the patented IsoYeast formulation, which includes isotopically labeled carbon and nitrogen sources as well as several other key chemical components, can improve the expression levels of the targeted proteins versus other defined, rich media. The media are also compatible with inclusion of various amino acids for selective amino acid labeling. No other yeast media currently offer this flexibility.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service